CPC (Colder) MPC32039 All-Plastic Quick Disconnect Sealing Cap, Polysulfone (PS), 5/pack
Representative image only.
CPC (Colder) Mfr # MPC32039 – Item # EW-31311-74
$34.75USD / Pkg of 5
BUY MORE & SAVE
  • 1-9$34.75USD / Pkg of 5
  • 10-49$31.28USD / Pkg of 5
  • 50-99$29.54USD / Pkg of 5
  • 100+$27.80USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeQuick-Disconnect
  • Min Temperature (° F)-40
  • Max Temperature (° F)300
  • Min Temperature (° C)-40
  • Max Temperature (° C)148
  • Max Operating Pressure (PSI)120
  • DescriptionAll-Plastic Quick Disconnect Sealing Cap, PS, 5/pack
Certificate of Compliance
More About this Item
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).