CPC (Colder) MPC3301239 Quick-disconnect body, polysulfone, 3/4" sanitary connector; MPC
Representative image only.
CPC (Colder) Mfr # MPC3301239 – Item # EW-31604-27
Attach directly to your sanitary hardware assemblies!
  • USP Class IV materials
  • Fully sterilizable
PRODUCT OPTIONS
Free Certificate of Comliance
$16.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodAutoclave, e-beam, ethylene oxide (ETO), or gamma
  • Connection TypeSanitary x Quick-Disconnect
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionQuick-disconnect body, polysulfone, 3/4" sanitary connector; MPC
Certificate of Compliance
Key Features
  • USP Class IV materials
  • Fully sterilizable
More About this Item
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies. All models feature USP Class VI platinum-cured silicone gaskets. Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements on page 509 for greater flexibility in your process system.