CPC (Colder) MPCK32003 Sealing Cap with Lock, Polycarbonate, 1/4", 5/pack
Representative image only.
CPC (Colder) Mfr # MPCK32003 – Item # EW-31311-67
$29.75USD / Pkg of 5
BUY MORE & SAVE
  • 1-9$29.75USD / Pkg of 5
  • 10-49$26.78USD / Pkg of 5
  • 50-99$25.29USD / Pkg of 5
  • 100+$23.80USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolycarbonate
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeQuick-Disconnect
  • Min Temperature (° F)-40
  • Max Temperature (° F)250
  • Min Temperature (° C)-40
  • Max Temperature (° C)121
  • Max Operating Pressure (PSI)125
  • DescriptionSealing Cap with Lock, Polycarbonate, 1/4", 5/pack
Certificate of Compliance
More About this Item
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).