CPC (Colder) NS4 Non-Spill Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, 1/4" ID
Representative image only.
CPC (Colder) Mfr # NS4D22004 – Item # EW-31603-40
Stop leaks in your line—spillage as low as 0.10mL
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation
PRODUCT OPTIONS
$15.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Seal MaterialEPDM
  • Valve Spring Material316 Stainless Steel
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • ColorGrey
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)120
  • DescriptionNS4 Non-Spill Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, 1/4" ID; 1/Ea
Key Features
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation
More About this Item
These CPC® non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. This non-spill series ensures minimal leaks and air inclusions in your process line—protects process purity and minimizes loss due to accidental disconnects. The NS4 and NS6 series non-spill couplings confirm secure engagement with an audible click.
The NS4 series offers a spillage rate of <0.1 mL. The NS6 series offers a spillage rate of <0.3 mL.
Choose one body and one insert, each from the same coupling series (NS4 or NS6) for a fitting that will work in multiple applications.