CPC (Colder) NS4D13006 Non-spill coupling body, polypropylene, 3/8" OD tubing compression fitting; 25/PK
Representative image only.
CPC (Colder) Mfr # NS4D13006 – Item # EW-31309-42
Less than 0.10mL spillage
  • One-hand operation
PRODUCT OPTIONS
$439.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeQuick-Disconnect x Compression
  • Compression Size / Tube Od (Port 1) (in)3/8
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)120
  • DescriptionNon-spill coupling body, polypropylene, 3/8" OD tubing compression fitting
Key Features
  • One-hand operation
More About this Item
Virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. For a complete fitting, order one body and one insert. All bodies will mate with inserts to make a fitting that will work for multiple applications.