CPC (Colder) Plastic Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Straight-Through, 1/2" ID; 5/Pk
Representative image only.
CPC (Colder) Mfr # MPX22803M – Item # EW-31311-63
Fittings connect in a snap
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
PRODUCT OPTIONS
Free Certificate of Comliance
$22.50USD / Pkg of 5
BUY MORE & SAVE
  • 1-9$22.50USD / Pkg of 5
  • 10-49$20.25USD / Pkg of 5
  • 50-99$19.13USD / Pkg of 5
  • 100+$18.00USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolycarbonate
  • Sterilization MethodAutoclave or gamma
  • Connection TypeQuick-Disconnect x Hose Barb
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • Quick-Disconnect Flow Size (in)1/2
  • Min Temperature (° F)-40
  • Max Temperature (° F)250
  • Min Temperature (° C)-40
  • Max Temperature (° C)121
  • Max Operating Pressure (PSI)60
  • DescriptionPlastic Quick-Disconnect Fitting, Hose Barb Insert, Polycarbonate, Straight-Through, 1/2" ID; 5/Pk
Certificate of Compliance
Key Features
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
More About this Item
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).