CPC (Colder) PLC14006 Quick-Disconnect Coupling Body, Push-to-Connect 3/8" OD; Straight; 25/PK
Representative image only.
CPC (Colder) Mfr # PLC14006 – Item # EW-31308-55
$338.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Seal MaterialBuna N
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x Push-to-Connect
  • Push-to-Connect Tube OD (port 1) (in)3/8
  • Quick-Disconnect Flow Size (in)1/4
  • Max Temperature (° F)180
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • Max Vacuum (in Hg)28
  • Max Vacuum (mm Hg)711
  • Length (cm)5.2832
  • DescriptionQuick-Disconnect Coupling Body, Push-to-Connect 3/8" OD; Straight
More About this Item
Quick-disconnect fitting with simple push-to-connect interface for your rigid tubing. Choose a compatible body and insert for a complete fitting system.