CPC (Colder) PLCD17004 Hose Barb Fitting Body; Valved, 1/4" Tube ID, 1/4" Flow Size, Acetal
Representative image only.
CPC (Colder) Mfr # PLCD17004 – Item # EW-06360-65
$11.20USD / Each
BUY MORE & SAVE
  • 1-9$11.20USD / Each
  • 10-49$10.08USD / Each
  • 50-99$9.52USD / Each
  • 100+$8.96USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Seal MaterialBuna N
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x Barb, Valved
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Quick-Disconnect Flow Size (in)1/4
  • Max Temperature (° F)180
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • Max Vacuum (in Hg)28
  • Max Vacuum (mm Hg)711
  • Length (cm)4.953
  • DescriptionHose Barb Fitting Body; Valved, 1/4" Tube ID, 1/4" Flow Size, Acetal
More About this Item
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.