CPC (Colder) PLCD40006 Quick-Disconnect Panel-Mount Compression Insert; Valved, 3/8" Tube OD; 25/PK
Representative image only.
CPC (Colder) Mfr # PLCD40006 – Item # EW-31307-54
$341.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Seal MaterialBuna N
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeQuick-Disconnect x Compression
  • Compression Size / Tube Od (Port 1) (in)3/8
  • Quick-Disconnect Flow Size (in)1/4
  • Max Temperature (° F)180
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • Max Vacuum (in Hg)28
  • Max Vacuum (mm Hg)711
  • Length (cm)4.8514
  • DescriptionQuick-Disconnect Panel-Mount Compression Insert; Valved, 3/8" Tube OD
More About this Item
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.