CPC (Colder) Quick-Disconnect Fitting, Compression Body, Panel-Mount, Polypropylene, Valved, 1/4" OD
Representative image only.
CPC (Colder) Mfr # PMCD120412 – Item # EW-06364-05
No more struggling—just press thumb latch
  • Fittings connect in a snap
PRODUCT OPTIONS
$19.00USD / Each
BUY MORE & SAVE
  • 1-9$19.00USD / Each
  • 10-49$17.10USD / Each
  • 50-99$16.15USD / Each
  • 100+$15.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Seal MaterialEPDM
  • Valve Spring Material316 Stainless Steel
  • Sterilization MethodGamma
  • Connection TypeQuick-Disconnect x Compression
  • Compression Size / Tube Od (Port 1) (in)1/4
  • Quick-Disconnect Flow Size (in)1/8
  • Flow PatternValved
  • Max Temperature (° F)160
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)120
  • Max Vacuum (in Hg)28
  • Max Vacuum (mm Hg)711
  • DescriptionQuick-Disconnect Fitting, Compression Body, Panel-Mount, Polypropylene, Valved, 1/4" OD; 1/Ea
Key Features
  • Fittings connect in a snap
More About this Item
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).