CPC (Colder) Quick-Disconnect Fitting, Valved, In-Line Coupling, 3/8"; 1/Ea
Representative image only.
CPC (Colder) Mfr # APCD17006SH – Item # EW-49007-36
Fittings connect in a snap
  • No exposed metal parts
  • Protects against accidental disconnects
PRODUCT OPTIONS
$8.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Seal MaterialBuna-N
  • Valve Spring Material316 Stainless Steel
  • Connection TypeQuick-Disconnect x Hose Barb
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • Flow PatternValved
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • DescriptionQuick-Disconnect Fitting, Hose Barb Body, Acetal, Valved, 3/8" ID with Shroud; 1/Ea
Key Features
  • No exposed metal parts
  • Protects against accidental disconnects
More About this Item
These CPC® APC-Series quick-disconnect fittings, combine a compact design with durable acetal construction, making them an excellent choice for many applications. All fittings feature a 1/4” flow, plastic thumb latch, fewer moving parts, and a smooth contoured design to deliver ease-of-use and excellent flow in a compact size. Simply press the thumb latch to disconnect fitting halves. Choose from in-line compression and hose barb bodies which feature shrouded latches to provide further protection against accidental disconnect. The ferruleless PTF compression fittings do not require ferrules to achieve a secure connection, which makes them easier to use and reuse. PTF fittings are designed for semi-rigid tubing, i.e., polyethylene, nylon, polyurethane, etc.
A complete coupler requires a fitting body and an insert (order separately).
Minimun 25 pcs required to purchase