CPC (Colder) Quick-Disconnect Fitting with Automatic Shutoff Valve, Hosebarb Body, Polysulfone, 3/8" ID; 10/Pk
Representative image only.
CPC (Colder) Mfr # HFCD17639M – Item # EW-31313-02
Eliminate the need for pinch clamps
  • Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
  • Valves restrict flow when fitting halves are disconnected
  • Fitting body constructed of durable polysulfone
PRODUCT OPTIONS
$311.00USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$311.00USD / Pkg of 10
  • 10-49$279.90USD / Pkg of 10
  • 50-99$264.35USD / Pkg of 10
  • 100+$248.80USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Seal MaterialPlatinum-Cured Silicone
  • Valve Spring Material316 Stainless Steel
  • Sterilization MethodAutoclave or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • ColorAmber
  • Min Temperature (° F)-40
  • Max Temperature (° F)280
  • Min Temperature (° C)-40
  • Max Temperature (° C)138
  • Max Operating Pressure (PSI)125
  • DescriptionQuick-Disconnect Fitting with Automatic Shutoff Valve, Hose Barb Body, Polysulfone, 3/8" ID; 10/Pk
Key Features
  • Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
  • Valves restrict flow when fitting halves are disconnected
  • Fitting body constructed of durable polysulfone
More About this Item
These CPC® quick-disconnect fittings feature an automatic shutoff valve which closes off the flow path at disconnection, protecting valuable media while eliminating the need for pinch clamps. The thumb latch enables easy one-handed operation. An audible click is heard when the body and insert are coupled to confirm a secure, leak-free connection.
For a complete fitting, order both a body and insert (order separately).