CPC (Colder) SXM420112 Coupling Insert, Six-Line, Male, Polypropylene, 1/16" ID Tube; 10/PK
Representative image only.
CPC (Colder) Mfr # SXM420112 – Item # EW-31307-92
Save time -- with one coupling you can connect two, six, or ten lines
PRODUCT OPTIONS
$149.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/16
  • DescriptionCoupling Insert, Six-Line, Male, Polypropylene, 1/16" ID Tube
More About this Item
Colder has made it easy to handle multiple tubing lines in your process that require connection and reconnection! These multiline couplings simplify the manual steps in your process for quick transfers and minimize your down time. Twin Tube™ series: With two lines in one fitting, you can connect or disconnect with the push of a button. Each line is capable of independently handling anything from vacuum to 120 psi. The coupling allows rotation of both tubing lines to prevent kinking. The Twin Tube™ series is available with hose barb connections only. Sixtube™ series: With a single connector, control six different lines! An efficient thumblatch design allows quick disconnecting with optional shutoff.