CPS Products HP3 Premium Refrigerant Charge Hose, 3 ft/90 cm
Representative image only.
CPS Products Mfr # HP3 – Item # UX-10110-95
1/4" Standard charging hoses
  • All-weather flexibility and strength
  • Extra deep knurling on nuts for easy grip
  • Available with MicroSeal™ or ball valve fittings
  • Leak-proof eight-sided crimp
  • Blow-off grooves channel refrigerant to prevent spray back
  • 1 year warranty
PRODUCT OPTIONS
$62.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPremium Refrigerant Charge Hose, 3 ft/90 cm
  • Warranty1 year
1 Year Warranty
Key Features
  • All-weather flexibility and strength
  • Extra deep knurling on nuts for easy grip
  • Available with MicroSeal™ or ball valve fittings
  • Leak-proof eight-sided crimp
  • Blow-off grooves channel refrigerant to prevent spray back
  • 1 year warranty
More About this Item
The hose has 1/4" SAE fittings with a 45° bend on one end. It can operate at 750 psi working pressure with a burst pressure of 3000 psi.
1/4" Premium Hoses For All Refrigerants, including R-410A
  • All-weather flexibility and strength
  • 800 psi working pressure; 4000 psi burst
  • Environment friendly nylon permeation barrier meets SAE J2196
  • Compatible with CFC, HCFC and HFC refrigerants & blends
  • Blow-off grooves channel refrigerant to prevent spray back
  • Available with MicroSeal™ or ball valve fittings
  • 1/4" SAE fittings with 45° bend on one end
  • Leak-proof eight-sided crimp superior hose construction with ""double
    barrier protection""