CPS Products MBHP5E Black Max Professional A/C Manifold Set, 5' Valve Hoses; psi and F Gauges
Representative image only.
CPS Products Mfr # MBHP5E – Item # UX-10111-05
Professional-grade two-valve service manifolds engineered to test, charge, and recover refrigerant from HVAC/R systems
  • Anodized aluminum block bodies Advanced TripleSeal™ Piston Valve System Non-flutter metal cases gauges Easy-to-read refrigerant/pressure scales
  • 1 year warranty
$241.50USD / Each
$161.00 - $239.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Refrigerant Connections1/4" SAE 45° with 5-ft hose set
  • Pressure MediaR-22, R-404A, R-410A
  • DescriptionBlack Max Professional A/C Manifold Set, 5' Valve Hoses; psi and F Gauges
  • Warranty1 year
1 Year Warranty
Key Features
  • Anodized aluminum block bodies Advanced TripleSeal™ Piston Valve System Non-flutter metal cases gauges Easy-to-read refrigerant/pressure scales
  • 1 year warranty
More About this Item
Rugged metal-cased gauges are non-flutter, with convenient front calibration access ports and easy-to-read refrigerant/pressure scales.
  • All-in-one HVAC gauge with R-410A, R-22, and R-404A scales
  • High-pressure 5 ft/150 cm PRO-SET hoses with ball valve fittings on end