CR Magnetics CR9450-NPN Current Switch, NPN Output
Representative image only.
CR Magnetics Mfr # CR9450-NPN – Item # EW-68480-57
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Relay TypeNPN
  • DescriptionCurrent Switch, NPN Output
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The current switch series is a low cost, self powered, fixed setpoint. They are designed for applications that require an on-off indication of current flow.
Current levels above the guaranteed full-on level will turn the output to full on. The current switch is recommended only for applications where the continuous operation current is above the rated full on level of 350mA. Operaiton below this point will not drive the output device full-on and derate the output ratings.
The unit is available with a NPN or PNP output transistor for switching DC and a SCR output for switching AC. Connections can be made directly to items such as a PLC or electromechanical relay. Note that connections made directly to an inductive device such as an electromechanical relay will require a customer supplied clamping diode for DC operation or a snubber network for AC operation.