Crosstex Sani-Roll Sterilization Tubing, Paper/Plastic, 6 In X100 Ft
Representative image only.
Crosstex Mfr # SCT6 – Item # EW-10703-04
Convenient and economical way to package instruments
  • For use in steam, chemical vapor and EtO sterilizers
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)5
  • Length (in)1200
  • Width (cm)15
  • Length (cm)3050
  • DescriptionSani-Roll Sterilization Tubing, Paper/Plastic, 6 In X100 Ft
Key Features
  • For use in steam, chemical vapor and EtO sterilizers
More About this Item
Sani-Rolls have easy to read color process indicators to signal exposure to sterilization conditions. Blue tinted plastic aids in the detection of puncture holes caused by pointed/sharp instruments. Complete paper and film separation and a triple seal.