Crosstex Sterilization Indicator Tape; 1/2 In X 60 Yds, Case 36 Rolls.
Representative image only.
Crosstex Mfr # STS – Item # EW-10703-58
  • Indicates specific temperature has been achieved during sterilization cycle
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)1/2
  • Length (in)2160
  • Width (cm)1.3
  • Length (cm)5486
  • DescriptionSterilization Indicator Tape; 1/2 In X 60 Yds, Case 36 Rolls.
Key Features
  • Indicates specific temperature has been achieved during sterilization cycle
More About this Item
Color change to dark tan ndicates specific tempearture has been reached during sterilization cycle. Tape effectively seals sterilization tubing, autoclave bags, and CSR sterilization wraps.