Crowcon GASMAN-FL Single-gas Monitor, Lel (combustibles)

Representative image only.
Crowcon Mfr # GASMAN-FL – Item # EW-86550-06
Datalog and download up to 3000 hours with time and date stamp
  • Rechargeable lithium-ion battery for 12 hours of operation
  • Rugged housing is IP 65/67 rated for dust and water resistance
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Gases MeasuredCombustibles
  • Min Combustible Range (% LEL)0
  • Max Combustible Range (% LEL)100
  • Combustible Resolution (ppm)1% LEL
  • Combustible Alarm Set Points20% LEL
  • Alarm85 dB (audible), visible, vibrating
  • FunctionSTEL, TWA, peak hold, zero, low-battery indicator
  • Height (in)4
  • Width (in)2
  • Depth (in)1 1/4
  • DescriptionSingle-gas Monitor, Lel (combustibles)
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Rechargeable lithium-ion battery for 12 hours of operation
  • Rugged housing is IP 65/67 rated for dust and water resistance
  • 2 year warranty

More About this Item

Gasman monitor is available in a variety of toxic gas configurations to meet your individual application. Small size and light weight ensure the monitors are comfortable to wear all day long. Visible flashing LED, audible 85 dB, and vibrating alarms quickly alert the wearer to potential gas hazards. Data is recorded by the monitor and can be downloaded to a PC or printer using the adapter/PC interface (sold separately).
What's included: calibration cap, pocket clip, and rechargeable lithium-ion battery. (Order adapter/PC interface separately.)
Approval: UL and cUL Class I Div. 1 Groups A, B, C, D (pending), ATEX, CE

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat