Culture Bottle Roller Stacking Platform; Double
Representative image only.
Item # EW-36202-54
Save lab counter space by stacking rollers
  • Allows placement of additional roller inside incubator
  • Constructed of cast acrylic and aluminum
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCast Acrylic and Aluminum
  • Width (in)18
  • Height (in)18 3/4
  • Depth (in)16
  • DescriptionCulture Bottle Roller Stacking Platform; Double
Key Features
  • Allows placement of additional roller inside incubator
  • Constructed of cast acrylic and aluminum
More About this Item
This raised platform is designed to fit inside an incubator to allow for stacking of one or two Low Profile rollers inside.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting.
Accommodates 2 additional roller units to provide additional benchtop space, or to stack multiple units.