Cytocool II Tissue Freezing Spray, 11 oz Cans; 12/Cs
Representative image only.
Cytocool Mfr # 8323 – Item # EW-48802-10
Ideal for rapid hardening of difficult-to-cut tissue specimens.
$666.00USD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionII Tissue Freezing Spray, 11 oz Cans; 12/Cs
More About this Item
Prevent over-cooling of your specimen with Cytocool II tissue freezing spray. The integrated nozzle eliminates the need to attach a separate handle to each can for more efficient applications. Spray does not contain chlorofluoro-carbons (CFCs).