Davis Instruments 11 Mark 3 Lifeboat Astronomical Sextant
Representative image only.
Davis Instruments Mfr # 011 – Item # EW-68511-66
  • 1 year warranty
PRODUCT OPTIONS
$64.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMark 3 Lifeboat Astronomical Sextant
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A sextant is used to determine the angle between an astronomical object and the horizon for the purposes of celestial navigation.
Mark 3 Sextant
Sometimes referred to as a "lifeboat" sextant, the Mark 3 is an inexpensive training sextant. Yet this model has taken sailors around the world. It is full-sized and has sunshades, but no optical magnification to help you find dim celestial bodies.
*Available for sale in the United States and Canada only.