Davis Instruments 25 Mark 25 Sextant with Beam Converger™ mirror
Representative image only.
Davis Instruments Mfr # 025 – Item # EW-68511-68
  • 1 year warranty
PRODUCT OPTIONS
$487.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMark 25 Sextant with Beam Converger™ mirror
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A sextant is used to determine the angle between an astronomical object and the horizon for the purposes of celestial navigation.
The top of the line Davis plastic sextant, the Mark 25 features our Beam Converger mirror, sometimes referred to as a "full horizon mirror". A special coating is applied to high quality optical glass to allow the user to see through this mirror, yet still pick up images as faint as low-light stars. The Mark 25 is illuminated by a long-life LED and made of an upgraded plastic that is very stable.
*Available for sale in the United States and Canada only.