Davis Instruments 26 Mark 15 Sextant with Sunshades
Representative image only.
Davis Instruments Mfr # 026 – Item # EW-68511-67
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMark 15 Sextant with Sunshades
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A sextant is used to determine the angle between an astronomical object and the horizon for the purposes of celestial navigation.
Mark 15 Sextant - this model has many great features including seven large sunshades, a 3 mm x 27 mm star scope, and easy-to-read micrometer drum vernier scale that reads to 2/10 of a minute. The Mark 15 has a 7" (18 cm) frame radius graduated from 120 degrees to -5 degrees and comes with a traditional half-silvered mirror.
*Available for sale in the United States and Canada only.