Davis Instruments 6312 Additional Digital Display
Representative image only.
Davis Instruments Mfr # 6312 – Item # EW-86403-62
Optimize performance of your Davis Instruments weather station
  • Compatible with different Davis Instruments weather stations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Wind Speed (mph)2
  • Max Wind Speed (mph)150
  • DescriptionAdditional Digital Display
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with different Davis Instruments weather stations
  • 1 year warranty
More About this Item
Get the most from your Davis Instruments weather stations with these accessories and components. Ensure the accessory is compatible with the weather station you have. Weather stations sold separately.
The console displays and records your station's weather data, provides graph and alarm functions, and interfaces to a computer using our optional WeatherLink® software.
Cabled Systems A cabled Vantage Pro2 station transmits outside sensor data from the Integrated Sensor Suite (ISS) to the console using a straight-through four-conductor cable.
Wireless Systems A wireless Vantage Pro2 station transmits outside sensor data from the ISS to the console via a low-power radio.
Available for sale in the United States and Canada only.