Davis Instruments 6372 Wireless Temperature Station
Representative image only.
Davis Instruments Mfr # 6372 – Item # EW-32245-44
Add-on sensors provide accurate weather montioring
  • Stainless steel temperature probe with 12 ft (3.6 m) cable
  • Transmitter
  • Battery inside weather-resistant shelter
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (Outside) (° F)150
  • Min Temperature (Outside) (° F)-40
  • Max Temperature (Outside) (° C)65
  • Min Temperature (Outside) (° C)-40
  • DescriptionWireless Temperature Station
Key Features
  • Stainless steel temperature probe with 12 ft (3.6 m) cable
  • Transmitter
  • Battery inside weather-resistant shelter
More About this Item
Get accurate, reliable weather monitoring from these quality add-on sensors.
Battery powered station includes stainless steel temperature probe with 12ft (3.6 m) cable, transmitter and battery inside weather-resistant shelter, and mounting hardware. Lithium battery lasts up to one year under normal conditions.
Data from up to eight stations may be viewed on a single Vantage Pro2 Console/Receiver (sold separately). Or, for a complete set of weather data, use with a Wireless Vantage Pro2 or Vantage Pro2 Plus. (You can add up to seven Wireless Temperature Stations to either of these.)
Compatible with Vantage Pro2, Vantage Vue; not compatible with original Vantage Pro.
Available for sale in the United States and Canada only.