DEARDORFF FITZSIMMONS CORP 77012 Bleach Refill, 12 11-oz. bleach cartridges
Representative image only.
DEARDORFF FITZSIMMONS CORP Mfr # 77012 – Item # EW-18105-02
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Eliminates hazard of bleach spills on skin or surfaces
SPECS & DESCRIPTION
Specifications & Description
  • FormatSpray
  • DescriptionBleach Refill, 12 11-oz. bleach cartridges
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Eliminates hazard of bleach spills on skin or surfaces
More About this Item
This two-bottle fixed-ratio trigger sprayer automatically mixes and dilutes 5.25% bleach concentrate with tap water to make a 5000 ppm bleach disinfectant solution for use on hard, non-porous surfaces. The sprayer keeps the bleach separate from the water until you spray, keeping the 10% solution fresh and effective. Refill the water cartridge with tap water and lock in the disposable bleach cartridges and system is ready for use. Item 18105-00 kit includes six, 11-oz. bleach cartridges, two, 5000 ppm sprayer heads, and two 11-oz. water bottles.
Not available for sale into California Not available for sale outside of the United States