This item is now discontinued. Please browse similar items
Decon Labs Bacdown 7106 Gel No-Rinse Skin Cleanser, 16 oz, cs/6
Representative image only.
Decon Labs Mfr # 7106 – Item # EW-86115-16
Nondrying skin cleanser can be used anywhere soap and water are not available
  • Gentle emollients soothe and moisten skin
  • Reduces bacteria count on hands prior to gloving or entering aseptic environments
SPECS & DESCRIPTION
Specifications & Description
  • FormatPump
  • Volume (Liters)0.5
  • DescriptionGel No-Rinse Skin Cleanser, 16 oz, cs/6
Key Features
  • Gentle emollients soothe and moisten skin
  • Reduces bacteria count on hands prior to gloving or entering aseptic environments
More About this Item
Special formula of 62% ethyl alcohol. Meets OSHA requirements for hand washing.
Call Us At1-847-549-7600
Live chat