Decon Labs Conflikt 4104 Disinfectant/Detergent, 1 Gallon
Representative image only.
Decon Labs Mfr # 4104 – Item # EW-86115-44
Ready-to-use disinfectant spray
  • Stable solution
  • USDA approved
  • EPA registered.
  • Refill bottle
PRODUCT OPTIONS
$93.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • FormatRefill
  • Volume (gallons)1
  • Volume (Liters)3.8
  • DescriptionDisinfectant/Detergent; 1 gallon
Key Features
  • Stable solution
  • USDA approved
  • EPA registered.
  • Refill bottle
More About this Item
This ready-to-use, prediluted quaternary ammonium disinfectant is ideal for cleaning and disinfecting hard surfaces in the lab, hospital, or just about anywhere that thorough disinfection efficacy is required. Effective against viruses, bacteria and fungi, HIV-1, E-coli and other pathogenic organisms including TB.
Not available for sale into California Not available for sale in Canada,