DeFelsko PosiTest ATM20 Adhesion Tester Dgtl 3000psi 0/20MPa
Representative image only.
DeFelsko Mfr # ATM20A – Item # EW-59784-36
Measures adhesion of coatings to metal, wood, concrete, and other substrates
  • Pull rate indicator manually monitors and adjusts the rate of pull
  • Requires no external power source—ideal for lab and field use
  • Large, easy-to-read LCD displays pressure value in psi or Mpa
  • Easily select dolly sizes, change measurement units or store readings with the touch of a button
  • Inexpensive, single-use dollies eliminate cleaning for re-use and can be kept as a permanent record
  • Self-aligning aluminum dolly enables accurate measurements on smooth or uneven surfaces
  • Environmentally sealed enclosure meets IP65 for weatherproof, dustproof, and shockproof
  • Quality hydraulic pump can be used in any position
  • Internal memory stores maximum pull-off pressure, rate of pull, test duration and dolly size for up to 200 pulls
  • Compatible with PosiSoft Desktop for viewing, printing, and storing measurement data
  • 1 year warranty
$2,195.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdhesion Tester Dgtl 3000psi 0/20MPa
  • Warranty1 year
1 Year Warranty
Key Features
  • Pull rate indicator manually monitors and adjusts the rate of pull
  • Requires no external power source—ideal for lab and field use
  • Large, easy-to-read LCD displays pressure value in psi or Mpa
  • Easily select dolly sizes, change measurement units or store readings with the touch of a button
  • Inexpensive, single-use dollies eliminate cleaning for re-use and can be kept as a permanent record
  • Self-aligning aluminum dolly enables accurate measurements on smooth or uneven surfaces
  • Environmentally sealed enclosure meets IP65 for weatherproof, dustproof, and shockproof
  • Quality hydraulic pump can be used in any position
  • Internal memory stores maximum pull-off pressure, rate of pull, test duration and dolly size for up to 200 pulls
  • Compatible with PosiSoft Desktop for viewing, printing, and storing measurement data
  • 1 year warranty
More About this Item
The portable, hand-operated adhesion tester is easy to use and ideal for measuring adhesion of coatings to metal, wood, concrete, and other substrates. The manual hydraulic pump is designed to apply smooth and continuous pressure with a single stroke. The pull rate indicator allows you to easily monitor and adjust the rate of pull in accordance with international test methods.
The tester is calibrated and certified to ± 1% accuracy using a NIST traceable load cell. It conforms to international standards including ASTM C1583/D4541/D7234/D7522, ISO 4642/16276-1, BS EN 12004-2, AS/NZS 1580.408.5, and others.