DeFelsko FM PosiTest Mechanical Coating Thickness Gauge; 0-80 mils
Representative image only.
DeFelsko Mfr # FM – Item # EW-59784-25
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 80 mil
  • DescriptionPosiTest Mechanical Coating Thickness Gauge; 0-80 mils
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
DF50201
For hot dip galvanizing, hard chrome, metalizing, paint, enamel, plastic coating, on steel
DF50202
For electroplating, thin paint films, phosphating, on steel
FEATURES
  • Positive visual and audible indicators to designate when thickness reading is established
  • "V" grooves in probe housing and instrument base to allow correct positioning on cylindrical objects
  • Go-No Go button can be preset for rapid measurement
  • New overall design, full supported, positive positioning, no pivoting tendencies during measurement
  • Refined magnetic and balanced arm assemblies, improved resolution and accuracy
  • Extra rugged housing, not affected by mechanical shock, water, acid or solvents
  • Probe contact and dial rotation all in one finger operation
  • Explosion proof, one year warranty