Degree Controls Flowmarker-Fog Fluid Canister Refill, 2.5 oz bottle
Representative image only.
Degree Controls Mfr # HDW-000130 – Item # EW-10322-38
Visualize natural air currents without creating disturbances
  • Nontoxic fog breaks down into carbon dioxide and water
  • Sufficient for about 2000 fog cycles of circa 3 seconds each
  • Pressurized can according to UN No.1950 “AEROSOLS”
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)73
  • Compatible With10322-30
  • DescriptionFlowmarker-Fog Fluid Canister Refill, 2.5 oz bottle
  • Warranty1 year
1 Year Warranty
Key Features
  • Nontoxic fog breaks down into carbon dioxide and water
  • Sufficient for about 2000 fog cycles of circa 3 seconds each
  • Pressurized can according to UN No.1950 “AEROSOLS”
  • 1 year warranty
More About this Item
The fog consists of FDA-compliant, nontoxic polyfunctional alcohol preparation, which meets standards for USP purity (food grade), and highly purified demineralized water.
Order fog generator separately as model 10322-30.