Delmhorst F-2000/H Digital Hops Moisture Meter

Representative image only.
Delmhorst Mfr # F-2000/H – Item # EW-59823-25
Don't let your crop spoil
  • Dual-range program measures moisture content of hops and alfalfa hay
  • Stores up to 100 readings and calculates the average for your convenience
  • Audible alarm warns you when the moisture content is higher than the set point
  • 1 year warranty

$395.00USD / Each
sign in for your price.
Include Electrodes Today!

SPECS & DESCRIPTION

Specifications & Description

  • Measurement TypePin
  • Temperature Compensation8
  • Display TypeDigital
  • AlarmYes
  • BatteryOne 9V (included)
  • Height (in)3
  • Depth (in)1 5/8
  • DescriptionDigital Hops Moisture Meter
  • Warranty1 year
1 Year Warranty

Includes

one 9 V battery.

Key Features

  • Dual-range program measures moisture content of hops and alfalfa hay
  • Stores up to 100 readings and calculates the average for your convenience
  • Audible alarm warns you when the moisture content is higher than the set point
  • 1 year warranty

More About this Item

This moisture meter provides information necessary to ensure baling and curing don''t result in fungal damage or fire hazards. Built-in calibration check confirms readings are accurate.


Order electrode and handle separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat