Diba BenchMark™ Microbore Fixed Column Endpiece, 3 mm, 0.2 µm SS frit; 4/pk
Representative image only.
Diba Mfr # 006BFE-03-SS – Item # EW-11941-92
Frit comes pre-assembled with each endpiece
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 0.2 µm diameter/pore
$343.00USD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsSS
  • Description0.2 µm
Key Features
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 0.2 µm diameter/pore
More About this Item
The endpieces make the primary seal to glass microbore columns.