Diba BenchMark™ Microbore Fixed Column Endpiece, 3 mm, 30 µm PTFE frit; 4/pk
Representative image only.
Diba Mfr # 006BFE-03-30 – Item # EW-11941-90
Frit comes pre-assembled with each endpiece
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 30 µm diameter/pore
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPTFE
  • Description30 µm
Key Features
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 30 µm diameter/pore
More About this Item
The endpieces make the primary seal to glass microbore columns.