Diba Chromatography Column Frit, 35 mm dia, 25 µm, PE; 20/pk
Representative image only.
Diba Mfr # 006FR-35-25-PE – Item # EW-11945-22
Select from a range of porosities for your media and chemistry
  • Extend the life of your columns with these replacement frits
PRODUCT OPTIONS
$204.00USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)35
  • Wetted MaterialsPE
  • Description25 µm
Key Features
  • Extend the life of your columns with these replacement frits
More About this Item
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit.