Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, M6 ISO Thread x 1/8" OD; 10/Pk
Representative image only.
Diba Mfr # 002314 – Item # EW-21940-03
Chemically inert flangeless fitting with permanently attached PTFE ferrule ideal for repeat connect/disconnect
  • Available in PTFE/SS or PTFE/PEEK material
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Available in PTFE/SS or PTFE/PEEK material
More About this Item
The Dibafit™ Gripper fitting system uses a one-piece ferrule which fits permanently onto the outside of semi-rigid tubing.