Dickson A791 ACC Pressure Kit; Washers/Fittings/Adapters
Representative image only.
Dickson Mfr # A791 – Item # EW-08410-80
Choose what you need for your data logger
  • Special fitting kits, software, and flash cards are available
PRODUCT OPTIONS
$236.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionACC Pressure Kit; Washers/Fittings/Adapters
Key Features
  • Special fitting kits, software, and flash cards are available
More About this Item
Update, upgrade, and customize your data collection system with software, cables, adapters, and other accessories.
Accessory kit includes one each of these fittings for the 1/4" NPT(M) pressure recorder connection: garden hose adapter; hose barb fitting; quick coupler female nipple; filter-type pulsation snubber; lever handle shut-off cock; copper tube flare fitting; polyethylene tube compression fitting; copper tube compression fitting.