Dickson 6" Circular Chart Recorder Paper, 0 to 100, No Units, 7 Days, 60/Pack
Representative image only.
Dickson Mfr # C657 – Item # EW-80009-12
Monitor with precision accuracy for quality control
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
PRODUCT OPTIONS
$76.50USD / Pkg of 60
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)6
  • Description6\" Circular Chart Recorder Paper, 0 to 100, No Units, 7 Days, 60/Pack
  • Warranty1 year
1 Year Warranty
Key Features
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
More About this Item
Choose the paper to meet your application needs.