Dickson R022 Pressure Filter Kit w/ Snubber; 10/20/25/30/40 micron
Representative image only.
Dickson Mfr # R022 – Item # EW-08410-84
Choose what you need for your data logger
  • Special fitting kits, software, and flash cards are available
PRODUCT OPTIONS
$42.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPressure Filter Kit w/ Snubber; 10/20/25/30/40 micron
Key Features
  • Special fitting kits, software, and flash cards are available
More About this Item
Update, upgrade, and customize your data collection system with software, cables, adapters, and other accessories.
Compatible With PW4 units, PR8 units, PR100, PR300, PR500, PR125, PR325, and PR525.
Includes Fittings: 10-micron filter (Air and Other Gases); 20-micron filter (Low Viscosity Fluids - under 30 SSU); 25-micron filter (Water and Light Oils - 30 to 225 SSU); 30-micron filter (Medium Type Oils - 225 to 500 SSU); 40-micron filter (Viscous Fluids - over 550 SSU)