Dickson SM325 Datalogger, large display, temperature 2-channel; Type K
Representative image only.
Dickson Mfr # SM325 – Item # EW-23036-20
Large 1-3/8'' tall display can be seen from across the lab — up to 25 feet away!
  • Up to 32,000 datapoints can be stored in memory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeTemperature (two external)
  • Max Temperature (° F)2000
  • Min Temperature (° F)-300
  • Min Temperature (° C)-184
  • Max Temperature (° C)1093
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)1.8
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)1
  • Memory Size32,000 data points
  • Battery Type4 x AA batteries
  • Display Type4-digit LCD, 1.375 in high digits
  • Length (in)4
  • Height (in)3
  • Width (in)1 1/2
  • DescriptionDatalogger, large display, temperature 2-channel; Type K
  • Warranty1 year
1 Year Warranty
Key Features
  • Up to 32,000 datapoints can be stored in memory
  • 1 year warranty
More About this Item
These dataloggers feature high/low alarms, min/max readings, delayed start times, and °F/C selections. Download data to your PC via USB, 9-pin serial port, or use the portable FLASH memory card. Software allows for simple viewing and zooming of logged data in a graphical or tabular (numeric table) format, order separately.
Optional 21CFR11 software meets pharmaceutical requirements including, password protection, electronic signatures, encrypted data in secured files and auditable trails to identify users, date & times and actions taken.