Dickson SW400 Data Logger Calibration Software, Single-Point
Representative image only.
Dickson Mfr # SW400 – Item # EW-18002-08
Monitoring walk-in freezers, coolers, HVAC systems, clean rooms, and storage rooms
  • Jumbo 4.5” x 3.4” screen
  • Probe model available (FH325)
  • User-defined display settings
  • 32KB storage
  • Audio/visual alarms
  • 1 year warranty
$297.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Logger Calibration Software, Single-Point
  • Warranty1 year
1 Year Warranty
Key Features
  • Jumbo 4.5” x 3.4” screen
  • Probe model available (FH325)
  • User-defined display settings
  • 32KB storage
  • Audio/visual alarms
  • 1 year warranty
More About this Item
Dickson’s new Flat Panel LCD Recorder lets you view detailed temperature (FT300/325/420) or temperature and humidity (FH320/325) data on its large screen and allows for optional PC downloads. Its large 4.5” x 3.4” display shows you current temperature and humidity readings and displays a graph of data ranging from 1 hour to 32 days. No paper charts or running back to your PC to see data, this recorder shows you everything at the push of a button.
However, if you would like to retrieve the data, the one-step data retrieval from datalogger to a FLASH memory card makes the routine of gathering data fast and easy. By pressing a single button, all your data will be downloaded to the removable memory card (optional), thereby eliminating the need to bring a laptop or move the datalogger. A Card Reader Kit, 68511-92, and the memory cards, 68511-91, are all you need for this data retrieval system.