Digestion Tubes, Volumetric, Glass, 100 mL; 50/Pk 78870-76
Representative image only.
Mfr # 4003006 – Item # EW-78870-76
  • Perform digestions of aqueous samples for metals analysis by Atomic Absorption (AA) or Inductively Coupled Plasma (ICP)
  • Conduct Kjeldahl digestion of food products or plant tissue for nitrogen & phosphorous analysis
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigestion Tubes, Volumetric, Glass, 100 mL; 50/Pk 78870-76
  • Warranty1 year
1 Year Warranty
Key Features
  • Perform digestions of aqueous samples for metals analysis by Atomic Absorption (AA) or Inductively Coupled Plasma (ICP)
  • Conduct Kjeldahl digestion of food products or plant tissue for nitrogen & phosphorous analysis