Digi-Sense 2" Dual-Scale Thermometers; 50-550F/10-290C
Representative image only.
Digi-Sense Item # EW-08103-30
304 SS dial case and stem
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)50
  • Min Temperature (° C)10
  • Max Temperature (° F)550
  • Max Temperature (° C)290
  • Accuracy±1% full-scale
  • Division (° C)5
  • Division (° F)5
  • Dial Size (in)2
  • Stem Diameter (in)0.142
  • Description2\" Dual-Scale Thermometers; 50-550F/10-290C
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
External adjustment nut allows for simple field calibration. Each thermometer features accuracy to 1%, stainless steel case and stem. Individually packaged with a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Test.
44mm (1.75”) and 50mm (2”) dial sizes feature a beaker clip with built in adjustment tool in place of the probe cover with pocket clip.