Digi-Sense 3-Wire RTD, Aluminum Head, 6" L, -328 to 1112F
Representative image only.
Digi-Sense Item # EW-90445-61
  • 316 Stainless steel sheath material
  • 1/2'' X 1/2'' NPT stainless steel hex nipple for head mounting fittings
  • Aluminum screw cover connection head
  • Low range or high range wire wound elements
  • .00385º C temperature coefficient
PRODUCT OPTIONS
$275.75USD / Each
$189.00 - $279.00USD / Each
for an additional
$189.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (")6
  • Probe TypeRTD
  • Connector Type1/2" NPT(M)
  • Sheath Length (in)6
  • Description3-Wire RTD, Aluminum Head, 6" L, -328 to 1112F
  • Warranty1 year
1 Year Warranty
Key Features
  • 316 Stainless steel sheath material
  • 1/2'' X 1/2'' NPT stainless steel hex nipple for head mounting fittings
  • Aluminum screw cover connection head
  • Low range or high range wire wound elements
  • .00385º C temperature coefficient
  • Custom built assemblies available upon request
  • 1 year warranty
More About this Item
These RTD assemblies consist of a single Low Range or High Range Wire Wound element constructed of 316 Stainless Steel sheath material, assembled together with an Aluminum screw cover connection head.