Digi-Sense Acorn Temperature 6 Thermometer, Pt 100 RTD
Representative image only.
Digi-Sense Item # EW-35626-20
This new thermometer offers the convenience and functionality of expensive meters at an economical price.
  • Splashproof keypad
  • HOLD button freezes measured readings
  • Switchable between °C and °F
  • MAX/MIN modes
  • Under-range and over-range error messages
  • Auto power-off 17 minutes after last key press
  • Nonvolatile memory retains calibration data and stored values even if battery is removed.
  • Optional protective rubber boot features a built-in stand.
  • Manual user calibration is a single point offset adjustment to any temperature within the range.
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-320
  • Max Temperature (° F)1562
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • Accuracy±0.4°F (±0.2°C); ±4.0°F (±2.0°C) above 199.9° and below -99.9°
  • Resolution0.1°
  • Low Battery IndicatorYes
  • Display Type0.875\" Display
  • Height (in)1 1/2
  • Width (in)5 1/2
  • Depth (in)2 3/4
  • Battery Life200 hours
  • BatteryAAA Battery (4)
  • DescriptionAcorn Temperature 6 Thermometer, Pt 100 RTD
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Splashproof keypad
  • HOLD button freezes measured readings
  • Switchable between °C and °F
  • MAX/MIN modes
  • Under-range and over-range error messages
  • Auto power-off 17 minutes after last key press
  • Nonvolatile memory retains calibration data and stored values even if battery is removed.
  • Optional protective rubber boot features a built-in stand.
  • Manual user calibration is a single point offset adjustment to any temperature within the range.
  • 3 year warranty
More About this Item
Accepts one RTD probe with Digi-Sense® three-pin connector sold separately.