Digi-Sense ASTM Like Liquid-In-Glass Thermometer; 54C / Congealing Point, Total Immersion, 20 to 100.6C, Organic Liquid Fill
Representative image only.
Digi-Sense Item # EW-08009-30
Now you can perform ASTM method measurements without the worry of mercury contamination
  • Designed for specific ASTM (American Society for Testing and Materials) test methods
  • Nonmercury blue spirit liquid
  • 5 year warranty
PRODUCT OPTIONS
$185.75USD / Each
$219.50 - $292.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12 19/64
  • Min Temperature (° C)20
  • Max Temperature (° C)100.6
  • Accuracy±0.6°C
  • Division (° C)0.2
  • Thermometer FillBlue Spirit
  • Overall Length (in)12.3
  • Overall Length (mm)312
  • Immersion Depth (mm)312
  • Stem MaterialWhite-Back Glass
  • ASTM Number54C
  • DescriptionASTM Like Liquid-In-Glass Thermometer; 54C / Congealing Point, Total Immersion, 20 to 100.6C, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty
Key Features
  • Designed for specific ASTM (American Society for Testing and Materials) test methods
  • Nonmercury blue spirit liquid
  • 5 year warranty
More About this Item
These thermometers are precision manufactured to conform to ASTM E-1 specifications except for not being mercury filled. Designed for use in quality control laboratories or when mercury filled thermometers can not be used but where ASTM standards are still needed. ASTM established thermometer standards suited for special applications, and assigned an ASTM number according to each specification.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered.