Digi-Sense Blood Bank Verification Thermometer; -2 to 10C
Representative image only.
Digi-Sense Item # EW-90311-21
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • 1 year warranty
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")7 1/10
  • Min Temperature (° C)-2
  • Max Temperature (° C)10
  • Accuracy±0.4°C
  • Division (° C)0.1
  • Thermometer FillBlue Spirit
  • Bottle FillPropylene Glycol
  • Overall Length (in)7.1
  • Overall Length (mm)180
  • Stem MaterialWhite-Back Glass
  • DescriptionBlood Bank Verification Thermometer; -2 to 10C
  • Warranty1 year
1 Year Warranty
Key Features
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • 1 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.