Digi-Sense Blood Bank Verification Thermometer; -5 to 20C SKU 9031122
Representative image only.
Digi-Sense Item # EW-90311-22
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • 1 year warranty
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")4 29/32
  • Min Temperature (° C)-5
  • Max Temperature (° C)20
  • Accuracy±0.75°C
  • Division (° C)0.5
  • Thermometer FillBlue Spirit
  • Bottle FillPropylene Glycol
  • Overall Length (in)4.9
  • Overall Length (mm)125
  • Stem MaterialWhite-Back Glass
  • DescriptionBlood Bank Verification Thermometer; -5 to 20C
  • Warranty1 year
1 Year Warranty
Key Features
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • 1 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.