Cole-Parmer Compact Penetration Thermocouple Probe, 2.5" L, Grounded d; Type T
Representative image only.
Cole-Parmer Item # EW-08439-84
Penetration Probes Are Designed For Soft and Semi-Soft Materials
  • Response Time: 2 seconds or greater
  • Probe comes with PVC Handle
  • Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
PRODUCT OPTIONS
$56.00USD / Each
BUY MORE & SAVE
  • 1-5$56.00USD / Each
  • 6-11$50.40USD / Each
  • 12-23$47.60USD / Each
  • 24+$44.80USD / Each
$197.00 - $290.00USD / Each
for an additional
$197.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")2 1/2
  • Probe TypeT
  • Min Temperature (° F)-328
  • Max Temperature (° F)300
  • Min Temperature (° C)-200
  • Max Temperature (° C)148
  • Temperature Accuracy±3.6°F (±2.0°C), or ±0.4% of reading above 32°F (0°C); ±0.8% of reading below 32°F (0°C)
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath material316 Stainless Steel
  • Handle MaterialPVC
  • Sheath Length (in)2.5
  • DescriptionCompact Penetration Thermocouple Probe, 2.5\" L, Grnd; Type T
  • Warranty6 month
6 Month Warranty
Key Features
  • Response Time: 2 seconds or greater
  • Probe comes with PVC Handle
  • Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).